Stikit™ Gold Abrasive Sheet Rolls, 3M
Stikit™ Gold abrasive sheets features continuous lengths of A-weight paper, easily cut to size to greatly reduce product waste. Aluminum oxide mineral abrasive delivers a sharp cut and uniform finish during collision repair.
- Customizable Adhesive-backed sheet to blocks, file shoes or pads, helping reduce waste
- Aluminum oxide mineral provides fast cut and long abrasive life
- Anti-loading coating prevents dust and debris from clogging the cutting surface
These sheet rolls feature Stikit™ adhesive backing, making them exceptionally fast and easy to attach and remove using power or hand sanding equipment.
Frequently Bought Together