Edsyn™ Soldering Tip Cleaner, Jensen Tools
No-Wet Soldering Tip Cleaner
Catalog # 76242-486
Supplier: TESTEQUITY LLC TE
Supplier #: EA439-1
Jump to:
Product Details & Documents
Soft wire mesh design cleans tips without using water and will not reduce tip temperature as with wet sponges.
- Never needs water
Instead of using a wiping motion as with regular sponges, just thrust the iron tip straight into the mesh and remove.
Specifications
- Description:No-Wet Soldering Tip Cleaner