Solder Soakers™ Tip Cleaning Sponge with Multiple Holes, Easy Braid
76247-414
- Color:Yellow
- Descripción:Solder Soakers™ Tip Cleaning Sponge with Multiple Holes
- Dimensiones:2.6×2.6"
- Grosor:⅝"
- Referencia:76247-414
This sponge is exclusively designed for the electronics industry, the soaker removes oxides and excess solders.
- Provides extended tip life and precise soldering
: RoHS compliant.