To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Anchura:½"
- Color:Clear
- Longitud:33 m (36 yds.)
- Grosor:5.5 mil.
- Referencia:76245-814
- Core Size:76 mm (3")
The tape is developed with a high-performance acrylic adhesive on the face side and a silicone adhesive on the liner side of a 1 mil clear PET carrier.
- Double linered – Adhesive 350
- Ideal for attaching silicone rubber elastomers and foam
Features acrylic and silicone adhesives for specialized applications such as silicone gasketing. A thin polyester carrier provides dimensional stability and improved handling with ease of die cutting and lamination compared to adhesive transfer tapes.
It is excellent for demanding applications where silicone materials must be bonded to a range of substrates.