San Jamar® Element™ Lever Roll Towel Dispenser
76249-040
- Color:Transparent Black Pearl
- Descripción:Lever Roll Towel Dispenser
- Referencia:76249-040
- An x Prf x Al:12½ × 8½ ×12¾"
- Para ser utilizado con:Hardwound Roll Towels (8" wide, 8" diameter)
With a 33% smaller footprint than standard roll towel dispensers, the Element™ lever roll towel dispenser is ideal for dispensing in tight spaces.
- High capacity, accommodates a standard 800' 8" roll towel
- Ergonomic handle design for ease of use
- Impact-resistant plastic construction
This high-capacity, universal dispenser works with all roll paper qualities with any size core, including coreless, solid and center pull rolls. Simple and low maintenance, the Element™ has minimal internal parts and coordinates with both Classic and Oceans® family products.