Orders LinkContactUsLinkComponent
Symbol Labels, Brady
Symbol Labels, Brady
Labels, electrostatic, matte finish, Polyimide, EIA Standard 471 Symbol, Black on amber, 6.4×6.4 mm (0.25×0.25"), 0.0044"
Catalog # 76241-626
Supplier:   Brady Worldwide
Número de proveedor #:  SL-11-PK
Symbol Labels, Brady
Catalog # 76241-626
Supplier:  Brady Worldwide
Symbol Labels, Brady
Labels, electrostatic, matte finish, Polyimide, EIA Standard 471 Symbol, Black on amber, 6.4×6.4 mm (0.25×0.25"), 0.0044"
Price
$5,445.88
Log in Para disponibilidad
Paquete de 5,400
Product Details & Documents
The card mounted static awareness labels alert users of electronic devices to the possibility of electrostatic damage.
  • Heat resistant and with permanent adhesive
  • Abrasion, chemical and corrosion resistant
  • Humidity, UV and weather resistant
Symbols used conform to Military Standard 129J and EIA (Electronics Industries Association) Standard 471.
Especificaciones
  • Color:
    Black on amber
  • Description:
    Labels, electrostatic, matte finish
  • Dimensions:
    6.4×6.4 mm (0.25×0.25")
  • Material:
    Polyimide
  • Thickness:
    0.0044"
  • Legend:
    EIA Standard 471 Symbol