KLEENEX® Facial Tissues, Kimberly Clark
Catalog # 76243-638
Supplier: KIMBERLY CLARK CORP
Specifications
- Color:White
- Descripción:Kleenex® Facial Tissues
- Referencia:76243-638
- Empaquetado:125 Sheets/Box, 1500 Sheets/Case
- Ply:2
Specifications
About this item
These tissues offer superior performance, a premium feel and essential softness. Popular choice in healthcare and medical offices, hotels and lodging and businesses in general.
Each Kleenex® tissue is 2-ply, absorbent, soft and strong
Each tissue features smart innovations like an ultra-soft feel
These bulk tissues are Forest Stewardship Council (FSC) certified
The signal sheet keeps you ahead of run-outs (final 10 tissues are cream instead of white, so able to know it's time to replace the box).
Unit size: 8.3" x 7.8"
Certifications: Forest Stewardship Council (FSC)
Packaging: Convenience case.