Rubber Stoppers, 1 Hole, United Scientific Supplies
Catalog # 470005-776
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Tamaño:6.5
- Referencia:470005-776
- Diámetro de la boca:34 mm
- Diámetro del fondo:27 mm
- No./lb.:16
- Nº de proveedor:RST6A-H
Specifications
About this item
Solid, black stoppers contain a high percentage of pure rubber and weigh less than neoprene.
- Resilient and pliable for a long time
- Low in free sulfur
- Can withstand adverse laboratory conditions without hardening
These black stoppers are made of solid natural rubber that stays pliable over a long period of time. Ideal for use with non-aggressive or concentrated solutions. Low sulfur content. Stoppers will withstand temperatures from –25 to 70 °C (–13 to 158 °F). Each stopper has its size marked in raised figures.
Packaging: Sold as one-pound bags.