Orders LinkContactUsLinkComponent
Differential Counters
Differential Counters
Supplier:   LW SCIENTIFIC SE
Differential Counters
Supplier:  LW SCIENTIFIC SE
Certificates
Differential Counters
Differential Counters
2 Options Available Below
Product Details & Documents
Gear and lever reliability and feel.
  • Lightweight and portable
  • Right and left reset dials
  • Non-slip bottom
  • Bell alarm for 100th cell counted
Traditionally, differential counters have been manual "gear and lever" machines, and many lab techs are accustomed to the feel of the levered keys. We offer both 5-key and 8-key manual counters for those traditional techs to count Baso, Eosin, Myelo, Juven, Stab, Seg, Lymph, and Mono cells. These counters feature pictures and names of each cell for easier identification. These manual counters are economical, durable, and will provide years of use, even in the most demanding laboratory settings.
Product Family Options
Información del producto
  • Dimensions
    Availability
    Price
  • 470309-552
    5-Key Differential
    5-Key Differential
    9.75L×3.25W×2"H
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6,859.81
    $6,859.81
    Each (1UN)
       $6,859.81
    Supplier Number: CTL-DIFM-05KY
  • 470309-554
    8-Key Differential
    8-Key Differential
    12.25L×3.25W×2"H
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $7,704.10
    $7,704.10
    Each (1UN)
       $7,704.10
    Supplier Number: CTL-DIFM-08KY