Orders LinkContactUsLinkComponent
Ward's LiveSlides®
Ward's LiveSlides®
Supplier:   VWR International
Ward's LiveSlides®
Supplier:  VWR International
Certificates
Ward's LiveSlides®
Ward's LiveSlides®
2 Options Available Below
Product Details & Documents
Never before has it been so easy to view microorganisms, including bacteria, fungi, and protists.
  • Available as bulk pack of 50 slides only, or pack of 5 slides with transfer pipettes
  • Uniform depth of viewing well perfect for maintaining a shallow depth of field
  • Partitions help keep specimens in the viewing area
  • Polycarbonate material is safe for indoor or outdoor use
  • Can be used with traditional or digital microscopes
Each LiveSlide® consists of a base slide, and a set of coverslips of different thicknesses. One is very thin (0.13 mm), perfect for high magnification, and the second is a bit thicker (0.25 mm) for all applications. Each coverslip has a thin plastic film on each side for protection, one green and one clear. Remove both films by either peeling the film off or using a small piece of tape on the corner and lifting. Discard the films.
Product Family Options
Información del producto
  • Availability
    Price
  • 470309-714
    LiveSlides® Bulk Pack
    LiveSlides® Bulk Pack
    Log in Para disponibilidad
    Log in Para disponibilidad
    Caja de 50
    Caja de 50 - $4,299.59
    $4,299.59
    Caja de 50
       $4,299.59
  • 470313-232
    LiveSlides® Protist Study Set
    LiveSlides® Protist Study Set
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 5
    Paquete de 5 - $745.17
    $745.17
    Paquete de 5
       $745.17