Estwing Chisel Head Hammers
Supplier: ESTWING MFG CO SE
Jump to:
Acerca de este artículo
Ideal for use with sedimentary rocks.
- Availabile in different head weights
Patented nylon end caps now makes these hammers the most durable on the market. The head and handles are forged in one piece and are fully polished. These tools have unsurpassed balance and are double tempered for added strength.
2 Options Available Below
Product Details & Documents
Ideal for use with sedimentary rocks.
- Availabile in different head weights
Patented nylon end caps now makes these hammers the most durable on the market. The head and handles are forged in one piece and are fully polished. These tools have unsurpassed balance and are double tempered for added strength.
Product Family Options
Información del producto
- LengthAvailabilityPrice
- 470309-728Chisel Hammer, 16 oz.Supplier Number: E3-16BLC
- 470309-720Chisel Hammer, 20 oz.Supplier Number: E3-20BLC
- 470309-722Chisel Hammer, 24 oz.Chisel Hammer, 24 oz.11.25"Discontinued-Supplier Number: -