Headspace Caps, MicroSolv
Crimp Cap, 20 mm, 10 mm Hole, Aluminum, Silicone/PTFE White/White 45° Shore A, 3.0 mm, Silver
Catalog # 97054-954
Supplier: MICROSOLV TECH CORP
Número de proveedor #: 95025-08-1S
Jump to:
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Especificaciones
- Septum:Silicone/PTFE White/White 45° Shore A, 3.0 mm
- Color:Silver
- Description:Crimp Cap
- Material:Aluminum
- Size:20 mm, 10 mm Hole