Septa for 8-425 Screw-Thread Vial Caps, Thermo Scientific
Proveedor: Thermo Scientific
Certificados
Acerca de este artículo
Septa for 8-425 screw thread caps are constructed of PTFE, PTFE/rubber, or PTFE/silicone to suit a variety of HPLC, GC and IC applications.
- PTFE Disk septa are chemically inert
- PFTE red rubber septa are economical for many GC applications
- PTFE/silicone septa are most popular for HPLC and IC applications
- Pre-slit septa are easier to pierce with thin gauge needles
Frequently Bought Together