ContactUsLinkComponent
Inserts for Standard Opening Crimp-Top Vial, 11 mm, Thermo Scientific
  66064-264
undefined
Inserts for Standard Opening Crimp-Top Vial, 11 mm, Thermo Scientific
  66064-264
 :  C4012-529L
  • Tapa incluida:
    No
  • Capacidad:
    0.20 ml
  • Descripción:
    Precision point insert
  • Matriz:
    Borosilicate glass
  • Referencia:
    66064-264
  • Bottom Style:
    Conical
  • Nº de proveedor:
    C4012-529L
These inserts are made of clear, borosilicate glass.
These inserts are custom fitted for use with standard opening, 2 ml, 12×32 mm vials. Flat-bottom construction optimizes the depth of the vial.