ContactUsLinkComponent
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
  97055-430
undefined
AQR™ Caps for RSA™ Autosampler Vials, MicroSolv Technology Corporation
  97055-430
 :  9509S-10C-B-M
  • Tamaño de la tapa:
    9-425 mm
  • Cap Type:
    Screw cap
  • Septo:
    Fitted Ultra Pure Clear Silicone/PTFE
  • Matriz:
    PP
  • Referencia:
    97055-430
  • Cap Color:
    Light Blue
  • Nº de proveedor:
    9509S-10C-B-M