Orders LinkContactUsLinkComponent
Celestron UpClose Binoculars
Celestron UpClose Binoculars
Celestron UpClose Binoculars
Certificates
Celestron UpClose Binoculars
Celestron UpClose Binoculars
3 Options Available Below
Product Details & Documents
High-quality binoculars at an affordable price.
  • Multi-coated optics increase light transmission resulting in brighter and sharper images with high contrast levels
  • Ergonomic thumb indents and finger ridges for extra non-slip comfort and ease of use
  • Water-resistant
  • Aligned with GLOBE protocols
These binoculars are perfect for a variety of activities, such as bird watching, sporting events, nature and wildlife, hunting, travel and ventures, concerts and live events, and boating.
Delivery information: Binocular straps and soft carrying case included.
Product Family Options
Product Information
  • Magnification
    Availability
    Price
  • 470164-702
    UpClose G2 Zoom Porro Binocular
    UpClose G2 Zoom Porro Binocular
    10-30×
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $2,365.50
    $2,365.50
    Each (1UN)
       $2,365.50
    Supplier Number: 71260
  • 470164-700
    UpClose G2 Porro Binocular
    UpClose G2 Porro Binocular
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,419.40
    $1,419.40
    Each (1UN)
       $1,419.40
    Supplier Number: 71250
  • 470164-698
    UpClose G2 Roof Binocular
    UpClose G2 Roof Binocular
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $737.17
    $737.17
    Each (1UN)
       $737.17
    Supplier Number: 71230