Orders LinkContactUsLinkComponent
Ward's® Kick Net
Ward's® Kick Net
Supplier:   SHIVDIAL SUD & SONS
Ward's® Kick Net
Certificates
Ward's® Kick Net
Ward's® Kick Net
2 Options Available Below
Product Details & Documents
For drag style collecting.
  • Durable netting
  • Easily stored and dried
  • Simple assembly in field
  • Mesh size is 200 microns
  • Aligned with GLOBE protocols
The Ward's Kick Net is everything you could want in a student kick net. The square meter collecting area is large enough to collect sufficient samples, but small enough for a single student to manage if necessary, although two students are best. Each end is stabilized by a large pine pole that won’t collapse in a current. The base and sides of the net are framed in muslin sleeves, giving god support and net control.
Product Family Options
Net Bundle
Product Information
  • Length
    Width
    Availability
    Price
  • 470308-238
    Kick Net Set
    Kick Net Set
    115 cm
    122 cm
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,042.65
    $1,042.65
    Log in for availability
    Case de 10 (1UN)
    Case de 10 (1UN) - $1,136.49
    $1,136.49
    Each (1UN)
       $1,042.65
    Case de 10 (1UN)
       $1,136.49
    Supplier Number: B11235H
Replacement Parts
Product Information
  • Length
    Width
    Availability
    Price
  • 470308-226
    Kick Net
    Kick Net
    115 cm
    109 cm
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $497.67
    $497.67
    Log in for availability
    Case de 50 (1UN)
    Case de 50 (1UN) - $542.46
    $542.46
    Each (1UN)
       $497.67
    Case de 50 (1UN)
       $542.46
    Supplier Number: B11235H