Orders LinkContactUsLinkComponent
Ward's® Manual Pipette Controller
Ward's® Manual Pipette Controller
Supplier:   MTC BIO, INC.
Ward's® Manual Pipette Controller
Supplier:  MTC BIO, INC.
Certificates
Ward's® Manual Pipette Controller
Ward's® Manual Pipette Controller
2 Options Available Below
Product Details & Documents
Ward's® Manual pipette controller is the precise and easy to use manual pipette controller that's always ready to use.
  • For glass or plastic pipettes
  • Accommodates pipettes up to 100 ml
  • Easy, single lever control
  • Autoclavable lower body
  • Fingertip speed control
After attaching a pipette, one squeeze of its large rubber filling bulb is all it takes to start pipetting. There are no wires, no tubing, and no battery to recharge.
Begin aspirating by touching the EZ-Flow™ lever which is designed to precisely control the filling speed, based on how gently or firmly the lever is pressed.
A built-in replaceable filter ensures optimum flow rate and protects the controller from liquid penetration and overfill. A separate button provides a positive blow-out function when required.
Product Family Options
Información del producto
  • Availability
    Price
  • 470307-996
    Manual Pipette Controller
    Manual Pipette Controller
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,835.67
    $1,835.67
    Each (1UN)
       $1,835.67
    Supplier Number: P6420_WARD
  • 470307-998
    Replacement Filter, 0.45 μm
    Replacement Filter, 0.45 μm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 5
    Paquete de 5 - $484.38
    $484.38
    Paquete de 5
       $484.38
    Supplier Number: P6080-FI_WARD