Specifications
- Descripción:Tubing, 1/4" I.D., 3/8" O.D., Autoclavable
- Longitud:50' (15.2 m)
- Matriz:TPE
- Referencia:76207-640
Specifications
About this item
Replace outdated glass vacuum traps with a shatterproof, pre-assembled system.
- Comes ready-to-use, no assembly required
- Shatterproof bottles offer improved safety
- VersaCap eliminates tube tangling
- Easy to empty without detaching tubing
- Inline hydrophobic filter included
- Optional ultra-stable secondary container
- Optional overflow bottle for CDC compliance
The Vactrap™ 2 vacuum trap system adheres to the strictest CDC recommendations for laboratory safety in vacuum applications.
Ideal for protecting vacuum lines and users, Vactrap™ safely contains waste produced during supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap™ comes ready to use, and fully assembled. Simply attach to your vacuum source and begin your experiment.
Vactrap™ is available as a one- or two-bottle complete system with safety bin, or as a one-bottle assembly without a bin. For the ultimate performance, the two-bottle system with bin is recommended. Offering 3l of total storage, this option's second bottle offers the greatest safety assurance and overflow prevention.