Cellophane Roll
Supplier: HYGLOSS PRODUCTS, INC.
Jump to:
About this item
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- ColorAvailabilityPrice
- 470003-396Cellophane, 20×60" RollSupplier Number: 7606
- 470003-398Cellophane, 20×60" RollSupplier Number: 7608
- 470003-394Cellophane, 20×60" RollSupplier Number: 7602
- 470014-924Cellophane, 20×60" RollSupplier Number: 7603