Slotted Weight Set of 12, Aluminum
Catalog # 470304-046
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Slotted Weight Set of 12, Aluminum
- Referencia:470304-046
Specifications
About this item
High quality for the classroom.
- Simplified design
- All are easily removed/added
Includes a 50 g aluminum hanger and 12 aluminum weights, 9×20 g, 1×10 g, and 2×5 g.