Rotating Platform
Rotating Platform
Catalog # 470304-036
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: RTP001
Jump to:
Product Details & Documents
A classic and fun demonstration.
- An economical solution
An essential accessory for experiments with the popular Bicycle Wheel Gyroscope as well as for demonstrating the effect of moment of inertia on rotational motion (the spinning skater effect), this economical Rotating Plate spins freely due to its heavy duty ball bearing axle.
Documents
Especificaciones
- Description:Rotating Platform