Pendulum Investigation Kit
Pendulum Investigation Kit
Catalog # 470304-028
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: PNLKIT
Jump to:
Product Details & Documents
Understanding pendulum motion is fundamental to the study of vibrations and waves.
- Includes three different pendulum balls
- Uniquely design pendulum clamp holds three at once
This unique kit includes everything necessary for a full exploration of the basic features of pendulum motion in a complete but economical set.
Students can assess the principal factors of mass, length, and amplitude and even venture into a consideration of air resistance.
The individual components are of proven design - the drilled balls have a larger hole on one end than the other to allow supension at the center of mass, and the triple support features clamps that make adjusting the cord length precise and easy.
Documents
Specifications
- Description:Pendulum Investigation Kit