Plastic Labware Value Set
Plastic Labware Value Set
Catalog # 470304-026
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: PLSKIT2
Jump to:
Product Details & Documents
Economical Choice for the Classroom.
- All items in the set with the exception of wash bottles are autoclavable
Economical, and convenient starter kit or refresher set. Includes an assortment of popular plastic labware items.
Ordering information: 5 Beakers (50, 100, 250, 500 and 1000 ml), 7 Graduated Cylinders (10, 25, 50, 100, 250, 500 and 1000 ml), 4 Wash Bottles (Unitary, 500 ml), 6 Storage Bottles (Square, Wide Mouth, 500 ml), 3 Erlenmeyer Flasks (Wide Mouth, with cap, 250 ml), 1 Test Tube Rack (with 12 plastic tubes)
Documents
Especificaciones
- Description:Plastic Labware Value Set