Bleeding Art Tissue Paper
Tissue Paper, 20 assorted colors, Assorted, 50.8 cm (20"), 76.2 cm (30")
Catalog # 470149-674
Supplier: DIXON TICONDEROGA COMPANY
: P0058506
Jump to:
Product Details & Documents
This assortment includes a wide selection of rich, vibrant colors for any art project. Colors blend when moistened to create exciting artistic effects. Recyclable.
Especificaciones
- Color:Assorted
- Description:Tissue Paper, 20 assorted colors
- Length:76.2 cm (30")
- Width:50.8 cm (20")