Lab Tray
Lab Tray
Catalog # 470303-962
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: LTRAY1
Jump to:
Product Details & Documents
Ideal for containing small spills.
- Low design takes up less space but still contains spills
- Autoclavable
Flat polypropylene laboratory trays serve as a drip pan. Economical trays measuring 15×11.5" are versatile for general laboratory use.
Especificaciones
- Description:Lab Tray