Orders LinkContactUsLinkComponent
Equal Volume Density Cube Sets
Equal Volume Density Cube Sets
Equal Volume Density Cube Sets
Certificates
Equal Volume Density Cube Sets
Equal Volume Density Cube Sets
2 Options Available Below
Product Details & Documents
High quality density cubes for the classroom.
  • Ideal for density experiments and to explore the physical properties of different materials
  • Compare masses for equal volumes
  • Uniform, reliable dimensions
  • Activity manual/reference guide included
Compare the masses and densities of these uniform 1" cubes. Explore the principles of bouyancy, and test the physical properties of metals and non-metals. Each set includes a reference table showing density values of individual specimens.
Ordering information: Set of 4 includes acrylic, pine, nylon, and aluminum, packaged in a poly bag.
Set of 10 includes acrylic, oak, pine, rosewood, steel, aluminum, copper, brass, nylon, and PVC, packaged in a wooden storage box.
Set of 12 includes acrylic, oak, pine, poplar, steel, aluminum, copper, brass, nylon, PVC, lignum vitae, and polypropylene, packaged in a plastic box.
Product Family Options
Product Information
  • Availability
    Price
  • 470014-710
    Density cube, set of 4
    Density cube, set of 4
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $300.60
    $300.60
    Each (1UN)
       $300.60
    Supplier Number: DCSET4
  • 470303-936
    Density cube, set of 10
    Density cube, set of 10
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,159.28
    $1,159.28
    Each (1UN)
       $1,159.28
    Supplier Number: DCSET10