Spouting Cylinder
Height: 195/16"
Catalog # 470303-934
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: CYSP01
Jump to:
Product Details & Documents
Demonstrate Torricelli’s theorem to show the relationship between water pressure and depth.
- Clear design
- Three nozzles at different heights
This clear acrylic cylinder includes three vertically aligned spouting nozzles positioned 43/4" apart.
Each spouting nozzle also includes a stopper so pressure at each height can be shown individually.
A filler hose barb at the top can be used to quickly refill the cylinder or maintain a constant water depth.
Especificaciones
- Dimensions:Height: 195/16"