Electric Calorimeter
Electric Calorimeter
Catalog # 470304-020
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: CLRE02
Jump to:
Product Details & Documents
A Standard and Electric Calorimeter in one.
- Electrical requirements are 6 V DC, up to 2A
This device can be used as a classic standard calorimeter or as an electric calorimeter. Made of aluminum and plastic, the calorimeter has a diameter of 10 cm, height of 11 cm, liquid capacity of 175 ml, and a heat capacity of 25 J/°C.
Documents
Especificaciones
- Description:Electric Calorimeter