Synthware Desiccators
7000 ml, 240 mm (top), 195 mm
Catalog # 76264-310
Supplier: KEMTECH AMERICA INC
Supplier #: B135124
Jump to:
Product Details & Documents
Our desiccators are made from high quality borosilicate glass.
- Heavy wall borosilicate construction
- Ceramic plate included
- Glass lid included
The desiccator is designed for operation under vacuum pressure and is great for desiccation of your samples or for storage of humidity sensitive materials.
Certifications: Meet ASTM standards.
Delivery information: Supplied with inner ceramic plate and glass lid.
Specifications
- Ø int.:240 mm (top)
- Volume:7000 ml
- Height:195 mm