Neutalizer, LancerAcid Rinse, LCA-A, Lancer
Catálogo # MSPP-LCA25A
Proveedor: GETINGE USA SALES, LLC. MS
Especificaciones
- Descripción:LancerAcid Rinse, LCA-A
- Tamaño:2×2.5 gal.
- Referencia:MSPP-LCA25A
Especificaciones
Acerca de este artículo
LCA-A is a acetic acid based neutralizer solution. Used as a rinsing agent.
Provides an excellent rinse on all standard laboratory soils and contaminants. Can be used in all glassware washers.