Cherokee Unisex Scrub Tops and Pants
Catalog # 470304-200
Supplier: DENLINE UNIFORMS INC BE
Specifications
- Talla de ropa:M
- Ajuste de la prenda:Regular
- Tipo:Inseam: 31"
- Aplicaciones de vestimenta:Medical
- Género:Unisex
- Color:Pewter
- Tela:65% polyester/35% cotton poplin
- Referencia:470304-200
Specifications
About this item
Reliable medical scrubs for every application.
- 65% polyester/35% cotton poplin
- Soil release fabric
These unisex scrubs are perfect for everyday medical and laboratory use. Unisex V-neck tops are available in 1 and 3 pockets design. Drawstring pants are available in regular, short, and tall lengths. Machine wash warm with similar colors. Use only non-chlorine bleach when needed. Tumble dry low. Medium iron if needed.
Ordering information: Multiple colors and additional sizes are available upon request.