Condensate Recirculators
Condensate Recirculating System, CRSY102, 115 V, 60 Hz, 5 A, 125 lbs.
27476-014
: CRSY102-1
These recirculators acts as a water delivery system for use with any Caron humidity controlled chamber.
- Environmentally friendly
- Dimensions: 22.8W×43.1L×17.7cm H (9×17×7")
- Durable powder coated metal as external material
- Water reservoir 7 gal.
- Water purity 100 to 125 KΩ.cm
These are designed to utilize tap water, eliminating the need to purchase more expensive water purification systems or maintain a supply of distilled or deionized water.
The single switch operation and automatic air purge minimize and simplify user interaction. The front/rear handles and casters provide easy mobility for installation and service.
The optional UV germicidal lamp deactivates microorganisms including bacteria, molds, protozoa, viruses and yeasts for contamination-free water.
- Model:CRSY102
- Description:Condensate Recirculating System
- Electrical:115 V, 60 Hz, 5 A
- Weight:125 lbs.
Services
- DescriptionDetails
- 76019-658PM kit for model CRSY102 condensate recirculatorPreventative maintenance kit for model CRSY102 (condensate recirculator) includes (1) CLP-130 SS clip, (1) DEM-103 demineralizer cartridge, (1) FIL-607 carbon cartridge, (1) HUM-120 copper mesh antimicrobial rod, (1) NOZ-110 atomizer, brass with mesh screen and (1) SIL-111 silicone grease. Detailed instructions included.KIT(1UN) - $13,056.40