Eppendorf® Pipette Holder System
Product Details & Documents
With space for up to six single- or multi-channel pipettes, users can easily store Eppendorf pipettes and multi-dispensers within easy reach with the Eppendorf pipette holder System. The flexible carousels, stands, and wall mount devices fit all current Eppendorf manual liquid handling instruments as well as most of their predecessors.
- Pipette Holder System stores all current mechanical and electronic Eppendorf pipettes (including the Repeater®) and most of their predecessors
- Pipette Carousel 2: rotatable holder with completely new design for all mechanical pipettes including Eppendorf Research®, Research plus, Reference, Reference 2, and Biomaster®
- Charger Carousel 2: carousel with intelligent charging electronics and increased capacity for up to six instruments (charges four instruments at a time and all six in turn)
- Carousels and stands with interchangeable pipette holders and charger shells, no tools required
- Large rubber feet protect carousels and stands from liquids spilled on bench top
- Pictograms on all charger shells and pipette holders for clear assignment in the lab
The robust Eppendorf pipette holder system allows users to store up to six Eppendorf pipettes or hand dispensers safely and securely in a small footprint, freeing up valuable bench space. Pipettes are positioned upright to prevent their nose cones from coming into contact with work surfaces and prevent damage and risk of cross-contamination. Pipette carousels and stands provide easy access to the instrument of choice, and are available with or without a charging station. Pipette stands carry one electronic pipette or Repeater®, and wall mount devices store one mechanical or electronic pipette, or Repeater®.
Ordering information: Includes charger shells for six Eppendorf Xplorer®/Xplorer® plus and holders for 6 Eppendorf Research®, Eppendorf Research® plus, Eppendorf Reference® 2.
Especificaciones
- Description:Charger stand 2 for one Repeater® E3/E3x
- Cat. No.:76204-686
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact