Buret Rack
Buret Rack, 12
Catalog # 470304-900
Supplier: EISCO SCIENTIFIC LLC
Supplier #: BURRCK12
Jump to:
Product Details & Documents
Safely store up to 12 burets.
- Natural wood
- Sturdy design
This buret rack accepts 12 burets of 50 mL capacity, up to 0.75" (2 cm) in diamater. The sturdy design allows burets to be fitted in and removed very easily. Optional brackets allow for the rack to be mounted to the wall. Rack dimensions are 32 x 3 x 23.5".
Specifications
- No. of Holes:12
- Description:Buret Rack