Beakers, Jacketed, Conical Bottom, Chemglass
Catálogo # 80066-216
Proveedor: CHEMGLASS INC.
Small Business Enterprise
Especificaciones
- Ext.Ø:60 mm
- Capacidad:50 ml
- Descripción:Jacketed Reaction Beaker, 37(I.D.)×67H mm
- Altura:77 mm
- Referencia:80066-216
- Nº de proveedor:CG-1103-01
Especificaciones
Acerca de este artículo
Designed for use in temperature controlled chemical and biological reactions.
Bottom interior surface is conical in shape to help eliminate spinout when used with egg shaped spinbars. Hose connections have an O.D. of 10 mm at the largest serration.