Mini Economy Splash Goggles
Perfect for elementary students with small faces.
- Direct and Indirect vent styles available
- Anti-fog coating available
- UV absorbing polycarbonate lens
Lightweight, green tinted PVC body gently conforms to the face, providing a secure and comfortable fit. Ideal for smaller faces and working in confined areas. Direct vent provides maximum air flow and protection against grinding, chipping, and cutting operations.
Frequently Bought Together