Specifications
- Talla de ropa:XL
- Aplicaciones de vestimenta:Laboratory
- Color:White
- Referencia:89207-872
- Chest Size:27 1/4"
- Inseam:29 1/2"
- Nº de proveedor:TY121SWHXL0025
- Sleeve:36 1/2"
Specifications
About this item
Tyvek® 400 garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
- Laydown collar
- Elastic wrists
- Attached skid-resistant boots
- Serged seams
Tyvek® 400 provides an ideal balance of protection, durability and comfort of any limited use fabric technology. Tyvek® 400 fabric offers an inherent barrier against particles (down to 1.0 micron in size). Protection is built into the fabric itself; there are no films or laminates to abrade or wear away. Tyvek® 400 fabric's durability advantage over microporous film fabrics delivers consistently better barrier, even after wear and abrasion. Applications include lead and asbestos abatement/remediation, general maintenance/operations, spray painting, general clean-up.
Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls feature laydown collar, elastic openings are wrists for a tighter fit, and attached Tyvek® FC boots,which have special coating to provide added skid resistance.