Orders LinkContactUsLinkComponent
VWR® Cleanroom Clogs
VWR® Cleanroom Clogs
VWR® Cleanroom Clogs
Supplier:  VWR International
Certificates
VWR® Cleanroom Clogs
VWR® Cleanroom Clogs
6 Options Available Below
Product Details & Documents
Cleanroom Clogs from VWR® provide comfort and durability for use in cleanroom environments.
  • Antistatic
  • Autoclavable
  • Smooth surface for day long comfort
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Non-slip sole
  • Anatomic heel basin
Cleanroom Clogs from VWR® have been designed with foot comfort in mind. Smooth surface and aeration holes on both sides provide day long comfort. Special conical design prevents spilled liquids from entering inside the clog.
These shoes are suitable for autoclave sterilization. They can also be cleaned in 100 °C with domestic detergents, liquid disinfecting solutions, EO, or formaldehyde.
Ideal for use with VWR® Cleanroom Disposable or Launderable Socks.
Ordering information: Clogs can be ordered by the pair or per case.
Packaging: Packaged six pair per case.
Product Family Options
Product Information
  • Shoe size
    Color
    Availability
    Price
  • XS (US: 4–5.5; EU: 35–36)
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $3,089.78
    $3,089.78
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $3,367.86
    $3,367.86
    Par
       $3,089.78
    Case de 6 (1UN)
       $3,367.86
  • S (US: 6–7.5; EU: 37–38)
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $3,089.78
    $3,089.78
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $3,367.86
    $3,367.86
    Par
       $3,089.78
    Case de 6 (1UN)
       $3,367.86
  • M (US: 8–9.5; EU: 39–40)
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $3,089.78
    $3,089.78
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $3,367.86
    $3,367.86
    Par
       $3,089.78
    Case de 6 (1UN)
       $3,367.86
  • L (US: 10–11.5; EU: 41–42)
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $3,089.78
    $3,089.78
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $3,367.86
    $3,367.86
    Par
       $3,089.78
    Case de 6 (1UN)
       $3,367.86
  • XL (US: 12–13.5; EU: 43–44)
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $3,089.78
    $3,089.78
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $3,367.86
    $3,367.86
    Par
       $3,089.78
    Case de 6 (1UN)
       $3,367.86
  • XXL (US: 14–15.5; EU: 45–46)
    White
    Log in for availability
    Log in for availability
    Log in for availability
    Par
    Par - $3,089.78
    $3,089.78
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $3,367.86
    $3,367.86
    Par
       $3,089.78
    Case de 6 (1UN)
       $3,367.86