Specifications
- Talla de ropa:XL
- Aplicaciones de vestimenta:Cleanroom,Laboratory
- Color:Yellow
- Referencia:10010-436
- Chest Size:271/4"
- Inseam:29"
- Nº de proveedor:QC127TYLXL000400
- Sleeve:361/2"
Specifications
About this item
A lightweight, and durable fabric, DuPont™ Tychem® 2000 utilizes the strength of DuPont™ Tyvek® fabric and a polyethylene coating.
- Standard fit hood
- Elastic wrists and ankles
- Serged seams
- Available NAFTA sourced
Tychem® 2000 fabric provides at least 30 minutes of protection against 42 chemical challenges. Tychem® 2000 is used for light splash protection in a variety of industrial environments, including petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing. Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing. Attached hood with elastic around face opening is standard fit. Storm flap covers zipper, which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper. Elastic openings at wrists and ankles allow for a tighter fit.