Orders LinkContactUsLinkComponent
Magnetic Mounting Sets
Magnetic Mounting Sets
Supplier:   MARKETLAB, INC.
Magnetic Mounting Sets
Supplier:  MARKETLAB, INC.
Certificates
Magnetic Mounting Sets
Magnetic Mounting Sets
2 Options Available Below
Product Details & Documents
Mount a dispenser on any magnetic surface.
  • Neoprene-coated magnets protect metal surfaces
  • Powerful, industrial-strength 30 lbs. magnets
Easily convert any existing wall mount unit with keyholes for use on metal surfaces with these exclusive magnetic mounting sets.
Product Family Options
Información del producto
  • Availability
    Price
  • 470236-392
    Magnetic Mount 2
    Magnetic Mount 2
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (2UN)
    Each (2UN) - $3,839.79
    $3,839.79
    Log in Para disponibilidad
    Paquete de 2
    Paquete de 2 - $5,574.45
    $5,574.45
    Each (2UN)
       $3,839.79
    Paquete de 2
       $5,574.45
    Supplier Number: 13991
  • 470236-394
    Magnetic Mount 4
    Magnetic Mount 4
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (4UN)
    Each (4UN) - $6,384.37
    $6,384.37
    Each (4UN)
       $6,384.37
    Supplier Number: 13992