Shotblocker
Shotblocker
Catalog # 470236-382
Supplier: MARKETLAB, INC.
Número de proveedor #: 23135
Jump to:
Product Details & Documents
Instantly block needle pain and reduce anxiety.
- Value-engineered Shot blocker immediately minimizes needle pain and anxiety
- Uses blunt contact points to saturate the sensory signals around the injection site
- Simple, non-invasive method creates an immediate distracting effect
- Single-use design reduces cross-contamination risks
- For intramuscular and subcutaneous injections including in the arm, hip, or thigh
- For adult and pediatric patients
Help students learn to cope with patients who have problems with needles. These single-use pads are designed with a multitude of blunt contact points. This activates nerves all around the injection point and helps prevent the patient from even noticing the injection.
Especificaciones
- Description:Shotblocker