ContactUsLinkComponent
Brady® SPC® Absorbents Re-Form™ Multiwik® Universal Absorbent Socks, Brady Worldwide
Brady® SPC® Absorbents Re-Form™ Multiwik® Universal Absorbent Socks, Brady Worldwide
Catalog # 76193-314
Supplier:  Brady Worldwide
undefined
Brady® SPC® Absorbents Re-Form™ Multiwik® Universal Absorbent Socks, Brady Worldwide
Catalog # 76193-314
Supplier:  Brady Worldwide
Supplier Number:  MS-50
Specifications
  • Descripción:
    Absorbent Socks
  • Dimensiones:
    3" Dia x 4' L
  • Referencia:
    76193-314
  • Absorción:
    40 gal.
Specifications
About this item
ReForm™ Multiwik® SOCs are dependable, flexible tubes that contain and absorb liquids, mold around corners, and conform to uneven surfaces.
  • Absorbency capacity: 40 gallons
  • Color: brown
  • Fill material: cellulose & recycled paper
  • Supplied 40 per case
Moldable SOCs wrap around machinery and conform to uneven surfaces to prevent leaks from reaching walkways. Highly versatile absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids. ReForm™ Multiwik® absorbents are universal economy SOCs that are wringable, incinerable, and absorb most non-aggressive fluids.