Plastic Well Slides
Plastic well slides, dia. 2-3 mm, depth 16 mm, 10 slides, 75×25 mm
Catalog # 470218-414
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: CSPL10
Jump to:
Product Details & Documents
Reusable slides for liquid or aquatic specimens.
- Quality slide-making supplies
- Hands-on scientific learning
Simply tear one off and use to view your favorite algae, protist, or small aquatic invertebrate organisms.
Packaging: These reusable slides are sold as a perforated sheet of 10.
Especificaciones
- Description:Plastic well slides, dia. 2-3 mm, depth 16 mm, 10 slides
- L×W:75×25 mm