Stirring Rod
Stirring Rod, Glass, 6"
Catalog # 470235-746
Supplier: GSC INTERNATIONAL SE
Número de proveedor #: 1900-6-GR
Jump to:
Product Details & Documents
Stirring rods made of borosilicate glass and with fire polished ends.
- Fire polished ends
- Sold as a gross of 144
- Made from borosilicate glass
Stir up your lab work with our 6” stir rods. These no-nonsense glass stirrers are made from durable borosilicate and feature fire-polished ends for a smooth, safe touch. Whether you're mixing solutions, compounds, or just giving your beaker a good swirl, these rods get the job done right. With a full gross of 144 rods, you'll have plenty on hand to tackle any lab task that comes your way. Stop fussing with flimsy, low-quality stirrers and upgrade to the reliable performance of our stir rods.
Caution: Not a toy. Use in a laboratory or educational setting only. Not for children 13 and under.
Especificaciones
- Description:Stirring Rod, Glass
- Length:6"