Macro Cuvettes, UV Quartz, Fireflysci
Cuvette, 3.500 ml, L×W 10×10 mm, L×W×H 12.5×12.5×45 mm, 10 mm, PTFE
Catalog # 76015-106
Supplier: FIREFLYSCI, INC.
Número de proveedor #: 1UV10
Jump to:
Product Details & Documents
Standard spectrophotometer cuvette for UV/VIS spectroscopy.
- Gives highly accurate results, only need to test sample once
- Comes in UV Quartz materials
- Excellent transmission values
- Fire-fused to ensure that cuvettes are robust and durable
- Two windows polished
Especificaciones
- Capacity:3.500 ml
- Description:Cuvette
- Lightpath:10 mm
- Cover Type:PTFE
- External Dimensions:L×W×H 12.5×12.5×45 mm
- Internal Dimensions:L×W 10×10 mm