Orders LinkContactUsLinkComponent
biOrb® Maintenance Materials
biOrb® Maintenance Materials
Supplier:   GEOGLOBAL PARTNERS
biOrb® Maintenance Materials
Supplier:  GEOGLOBAL PARTNERS
Certificates
biOrb® Maintenance Materials
biOrb® Maintenance Materials
2 Options Available Below
Product Details & Documents
Keep Your biOrb® Working Like New
  • Original maintenance materials
  • Maintenance for the inside and outside
These biOrb® maintenance materials are perfect for maintaining all biOrb® aquarium products. As regular maintenance all biOrb® aquaria require regular filter changes about every 4 to 8 weeks. The service kits contain all you need for normal filter changes, a new cartridge, a special cleaning pad for the interior of the aquarium, and instructions. The Marine service kit also includes some reef grade salt and water supplements to balance your water after a change. If you like to stock ahead, the Service pack X3 includes all you need to make 3 filter changes, and also includes a bottle of water optimizer.
Is the exterior of your biOrb® looking like it needs some help. The acrylic aquaria doesn't need much care, but over time you want to clean it up. In order to keep your biOrb® looking its best, the manufacturer recommends using these cleaners, polishes and cleaning cloths on the acrylic finish.
Documents
Product Family Options
Exterior
Información del producto
  • Availability
    Price
  • 470234-868
    Cleaner Pump Pack
    Cleaner Pump Pack
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $436.96
    $436.96
    Log in Para disponibilidad
    Case de 6 (1UN)
    Case de 6 (1UN) - $476.29
    $476.29
    Each (1UN)
       $436.96
    Case de 6 (1UN)
       $476.29
    Supplier Number: 46025
Water Treatment
Información del producto
  • Availability
    Price
  • 470234-860
    Water Optimizer
    Water Optimizer
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $364.39
    $364.39
    Log in Para disponibilidad
    Case de 6 (1UN)
    Case de 6 (1UN) - $397.18
    $397.18
    Each (1UN)
       $364.39
    Case de 6 (1UN)
       $397.18
    Supplier Number: 46020