Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cubos
Accessories for Compact Bucket System, Contec
Accessories for Compact Bucket System, Contec
Accessories for Compact Bucket System, Contec
76199-896
:
CONTEC, INC. MX
Accessories for Compact Bucket System, Contec
76199-896
:
CONTEC, INC. MX
:
UBKT0008
Capacidad:
15 L (3.9 gal.)
Color:
Assorted
Descripción:
Red, blue and gray polypropylene autoclavable buckets
Referencia:
76199-896