Orders LinkContactUsLinkComponent
PIPETMAN® Diamond Tips EasyPack™, Gilson®
PIPETMAN® Diamond Tips EasyPack™, Gilson®
D1000 Diamond EasyPack™, 100 - 1000 µl, 1000 Tips
Catalog # 76180-024
Supplier:   GILSON, INC.
Número de proveedor #:  F161671
PIPETMAN® Diamond Tips EasyPack™, Gilson®
Catalog # 76180-024
Supplier:  GILSON, INC.
PIPETMAN® Diamond Tips EasyPack™, Gilson®
PIPETMAN® Diamond Tips EasyPack™, Gilson®
D1000 Diamond EasyPack™, 100 - 1000 µl, 1000 Tips
Price
$1,817.55
Log in Para disponibilidad
Paquete de 1,000
Product Details & Documents
PIPETMAN® DIAMOND Tips are designed to fit your PIPETMAN pipettes perfectly. They deliver accurate and precise liquid volumes when used with Gilson pipettes. EASYPACK™ is an economical way to order tips in bulk.
  • Accurate and precise
  • Reliable
  • Economical
Using PIPETMAN DIAMOND Tips with your PIPETMAN ensures maximum accuracy and precision and delivers consistent results. The perfect air-tight seal prevents leaks. The tips are made of pure polypropylene to deliver the correct volume and ensure complete dispensing. Volume graduations make it easy to visually check the volume aspirated.
The tips are manufactured in a clean room environment ensuring they are free of detectable contaminants that can affect your results. From mold to bench, they are guaranteed, certified, and traceable to meet the most stringent demands in many applications. Ordering in bulk and simply reloading the tips into empty racks saves you money.
Documents
Especificaciones
  • Volume:
    100 - 1000 µl
  • Packed:
    1000 Tips
  • Tip Style:
    D1000 Diamond EasyPack™