Orders LinkContactUsLinkComponent
VWR® Cleanroom Socks
VWR® Cleanroom Socks
VWR® Cleanroom Socks
Supplier:  VWR International
Certificates
VWR® Cleanroom Socks
4 Options Available Below
Product Details & Documents
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments.
  • Cleanroom compatible
  • No particle generation
  • Launderable
Minimize the risk of particle contamination with Cleanroom Socks from VWR®
Ideal use with VWR® Cleanroom Clogs
Available in either launderable or disposable options, Cleanroom Socks provide comfort for extended wear and are cleanroom compatible.
 : Socks are packaged 1 pair per bag, 200 bags per case.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 76102-192
  • 76102-194
  • 76102-196
  • 76102-198
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Fabric:
  • 100% polyester
  • 100% polyester
  • 100% polyester
  • 100% polyester
Especificaciones
Product Family Options
Información del producto
  • Type
    Color
    Garment fit
    Shoe size
    Availability
    Price
  • VWR® Launderable cleanroom socks
    White
    Regular
    S
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Par
    Par - $145.10
    $145.10
    Log in Para disponibilidad
    Case de 200 (1UN)
    Case de 200 (1UN) - $158.16
    $158.16
    Par
       $145.10
    Case de 200 (1UN)
       $158.16
    Especificaciones:
    More
    Specifications
    Cat. No.
    76102-192
    Garment application
    Cleanroom
    Gender
    Unisex
    Fabric
    100% polyester
  • VWR® Launderable cleanroom socks
    White
    Regular
    M
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Par
    Par - $145.10
    $145.10
    Log in Para disponibilidad
    Case de 200 (1UN)
    Case de 200 (1UN) - $158.16
    $158.16
    Par
       $145.10
    Case de 200 (1UN)
       $158.16
    Especificaciones:
    More
    Specifications
    Cat. No.
    76102-194
    Garment application
    Cleanroom
    Gender
    Unisex
    Fabric
    100% polyester
  • VWR® Launderable cleanroom socks
    White
    Regular
    L
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Par
    Par - $145.10
    $145.10
    Log in Para disponibilidad
    Case de 200 (1UN)
    Case de 200 (1UN) - $158.16
    $158.16
    Par
       $145.10
    Case de 200 (1UN)
       $158.16
    Especificaciones:
    More
    Specifications
    Cat. No.
    76102-196
    Garment application
    Cleanroom
    Gender
    Unisex
    Fabric
    100% polyester
  • VWR® Launderable cleanroom socks
    White
    Regular
    XL
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Par
    Par - $145.10
    $145.10
    Log in Para disponibilidad
    Case de 200 (1UN)
    Case de 200 (1UN) - $158.16
    $158.16
    Par
       $145.10
    Case de 200 (1UN)
       $158.16
    Especificaciones:
    More
    Specifications
    Cat. No.
    76102-198
    Garment application
    Cleanroom
    Gender
    Unisex
    Fabric
    100% polyester